missing translation for 'onlineSavingsMsg'
Learn More

Sodium Selenite Pentahydrate, Honeywell Fluka™

Puriss. p.a.,≥99.0% (RT)

42.90 € - 129.76 €

Chemical Identifiers

CAS 26970-82-1
Molecular Formula H10Na2O8Se
Molecular Weight (g/mol) 263.023
MDL Number MFCD00149173
InChI Key TUANAMBRHOLYTH-UHFFFAOYSA-L
Synonym sodium selenite pentahydrate, disodium selenite pentahydrate, unii-0wv4l961zv, ccris 1378, na2seo3.5h2o, selenious acid, disodium salt, pentahydrate, disodium pentahydrate selenite, sodium selenite hydrate 2:1:5, sodium selenite hydrate, disodium selenite hydrate
PubChem CID 134930
IUPAC Name disodium;selenite;pentahydrate
SMILES O.O.O.O.O.[O-][Se](=O)[O-].[Na+].[Na+]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15670510
View Documents
Honeywell-Fluka
00163-25G
25 g
42.90 €
25g
Please to purchase this item. Need a web account? Register with us today!
15660510
View Documents
Honeywell-Fluka
00163-100G
100 g
129.76 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

26970-82-1
263.023
TUANAMBRHOLYTH-UHFFFAOYSA-L
134930
O.O.O.O.O.[O-][Se](=O)[O-].[Na+].[Na+]
H10Na2O8Se
MFCD00149173
sodium selenite pentahydrate, disodium selenite pentahydrate, unii-0wv4l961zv, ccris 1378, na2seo3.5h2o, selenious acid, disodium salt, pentahydrate, disodium pentahydrate selenite, sodium selenite hydrate 2:1:5, sodium selenite hydrate, disodium selenite hydrate
disodium;selenite;pentahydrate

Specifications

26970-82-1
Na2SeO3 · 5H2O
2630
TUANAMBRHOLYTH-UHFFFAOYSA-L
disodium;selenite;pentahydrate
134930
≥99% (RT)
H10Na2O8Se
MFCD00149173
sodium selenite pentahydrate, disodium selenite pentahydrate, unii-0wv4l961zv, ccris 1378, na2seo3.5h2o, selenious acid, disodium salt, pentahydrate, disodium pentahydrate selenite, sodium selenite hydrate 2:1:5, sodium selenite hydrate, disodium selenite hydrate
O.O.O.O.O.[O-][Se](=O)[O-].[Na+].[Na+]
263.023
263.01g/mol
Sodium Selenite Pentahydrate
Videos
Safety and Handling

Safety and Handling

P261-P273-P280-P301 + P310 + P330-P304 + P340 + P312-P403 + P233

H300-H317-H331-H411

missing translation for 'einecsNumber' : 233-267-9

missing translation for 'rtecsNumber' : VS7420000

Product Identifier
  • Sodium selenite pentahydrate

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 2

Hazard Statement
  • H300 + H330-Fatal if swallowed or if inhaled.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • EUH031-Contact with acids liberates toxic gas.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications