Learn More
Diversey Soft Care Sensitive
Fragrance-free, dye-free mild hand wash for sensitive skins. Proves a great solution for areas where frequent washing is required and healthy hands are desired.
Brand:  Diversey 101108201
 View more versions of this product
Product Code. 17853105
  • 40.90 € / 10 L
Estimated Shipment: 14-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Biodegradable and ethically sourced
  • Skin compatability tested (Patch test)
  • Eco-certified with Nordic Swan Ecolabel
  • Endorsed by Swedish Allergy and Asthma Foundation
  • Safe, quick and effective hand wash helps reduce the spread of germs
  • Effective surfactant system ensures excellent removal of grease and other soils
  • Wetting agents provide even application and an easy rinse
  • Reduced skin irritation and greater skin compatibility with a pH balanced formula
  • Moisturizes hands with softening emollients, maintaining skinhealth for frequent users
  • Rubs easily onto hands and leaves no sticky residue
Specifications
Specifications
Gentle Soap
Liquid
Yes
Non-sterile
Clear
Single-use
2 x 5 L
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Brochures
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Diversey Soft Care Sensitive > 2x5 L canister