missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Disposable Closed XRF X-Cell Sample Cups

Use these Disposable Closed XRF X-Cell Sample Cups for fast, dependable, and reproducible XRF analysis of liquid samples.

Brand:  Cole-Parmer™ 3565

 View more versions of this product

Product Code. 15320562

  • 107.00 € / Pack of 100

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Requires : Window film
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Horiba Sulfur Analyzers
1.7 in.
XRF Analysis
100 Pk.
Sample Cup
43 mm
Polyethylene
Closed
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cole-Parmer™ Disposable Closed XRF X-Cell Sample Cups > Disposable Closed XRF X-Cell Sample Cups; Diameter: 43 mm; Top Style: Closed

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.