Learn More
SRD5A1 293T Cell Overexpression Lysate (Denatured), Abnova
Brand:  Abnova H00006715-T01.100uL
Product Code. 16190962
  • 215.00 € / 100µL
Explore more special offers
Description
Description
Western Blotting
Specifications
Specifications
293T
SRD5A1 293T Cell Transient Overexpression Lysate(Denatured)
28.6
pCMV-SRD5A1 full-length
100 μL
Human
Western Blot
BC007033
6.8
Transient overexpression cell Lysate was tested with Anti-SRD5A1 antibody (H00006715-B01) by western blotting
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Show More
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
SRD5A1 293T Cell Overexpression Lysate (Denatured), Abnova > 100μL