Learn More
RETSCH Stainless Steel Grinding Jar
Use these grinding jars for dry, wet, and cryogenic grinding of small samples. Retsch™ Stainless Steel Grinding Jars feature a self-centering mechanism and a screw-top design for leak-proof grinding.
Brand:  RETSCH 01.462.0313
 View more versions of this product
Product Code. 15300974
  • 4005.00 € / Each
Estimated Shipment: 09-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Screw-top grinding jars
  • Available in a variety of sizes
  • For use with Retsch mills
Specifications
Specifications
Grinding Jar
125 mL
For EMAX
Stainless Steel
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
RETSCH Stainless Steel Grinding Jar > Capacity: 125 mL; For EMAX