Learn More
RETSCH Stainless Steel Grinding Jar
Use these grinding jars for dry, wet, and cryogenic grinding of small samples. Retsch™ Stainless Steel Grinding Jars feature a self-centering mechanism and a screw-top design for leak-proof grinding.
400.00 € - 4005.00 €
Product Code | Brand | Capacity (Metric) | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | For Use With (Equipment) | Price | Quantity & Availability | |||||
10504041
|
RETSCH
02.462.0057 |
1.5 mL | For MM 200 | N/A | ||||||
14821631
|
RETSCH
01.462.0331 |
10 mL | For CryoMill |
1350.00 €
Each |
Estimated Shipment: 30-06-2025
Log in to see stock. |
|||||
10099031
|
RETSCH
01.462.0230 |
1.5 mL | For MM 301 Mixer mill |
1320.00 €
Each |
Estimated Shipment: 30-06-2025
Log in to see stock. |
|||||
10034170
|
RETSCH
02.462.0061 |
10 mL | For MM 200 | N/A | ||||||
10334172
|
RETSCH
014620236 |
10 mL | For MM 301 Mixer mill |
1415.00 €
Each |
Estimated Shipment: 27-05-2025
Log in to see stock. |
|||||
15300974
|
RETSCH
01.462.0313 |
125 mL | For EMAX |
4005.00 €
Each |
Estimated Shipment: 30-06-2025
Log in to see stock. |
|||||
15370994
|
RETSCH
01.462.0333 |
35 mL | For CryoMill |
1535.00 €
Each |
Estimated Shipment: 30-06-2025
Log in to see stock. |
|||||
15370964
|
RETSCH
01.462.0305 |
50 mL | For EMAX |
2735.00 €
Each |
Estimated Shipment: 30-06-2025
Log in to see stock. |
|||||
10232672
|
RETSCH
01.462.0214 |
35 mL | For MM 301 Mixer mill |
1740.00 €
Each |
Estimated Shipment: 17-06-2025
Log in to see stock. |
|||||
10626275
|
RETSCH
01.462.0334 |
25 mL | For CryoMill |
1215.00 €
Each |
Estimated Shipment: 12-06-2025
Log in to see stock. |
|||||
10399770
|
RETSCH
02.462.0059 |
5 mL | For MM 200 | N/A | ||||||
10160572
|
RETSCH
01.462.0231 |
5 mL | For MM 301 Mixer mill |
1355.00 €
Each |
Estimated Shipment: 30-06-2025
Log in to see stock. |
|||||
Description
- Screw-top grinding jars
- Available in a variety of sizes
- For use with Retsch mills
Specifications
Grinding Jar | |
Stainless Steel |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
RETSCH Stainless Steel Grinding Jar