Learn More
RETSCH Stainless Steel Grinding Jar
Use these grinding jars for dry, wet, and cryogenic grinding of small samples. Retsch™ Stainless Steel Grinding Jars feature a self-centering mechanism and a screw-top design for leak-proof grinding.
Brand:  RETSCH 02.462.0059
 View more versions of this product
Product Code. 10399770
Description
Description
  • Screw-top grinding jars
  • Available in a variety of sizes
  • For use with Retsch mills
Specifications
Specifications
Grinding Jar
5 mL
For MM 200
Stainless Steel
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
RETSCH Stainless Steel Grinding Jar > Capacity: 5 mL; For MM 200