Learn More
Buerkle™ Stainless Steel Spatula
Universal instrument for collecting, dividing and mixing pastes, creams etc.
Brand: Buerkle™ 5313-0300
Description
Universal instrument for collecting, dividing and mixing pastes, creams etc.- Blade made of stainless steel V2A (1.4301)
- Wooden handle
Specifications
Spatula | |
Stainless Steel V2A | |
Stainless Steel Blade, Wood Handle |
Universal Instrument for Collecting, Dividing and Mixing Pastes, Creams | |
Wood | |
300 mm |
Product Suggestions
Customers who viewed this item also viewed
Videos
-
Buerkle Zone samplers Multi sampler, All-layer sampler, Uno sampler