Learn More
Buerkle™ Stainless Steel Spigot
Very sturdy and robust spigot made of stainless steel AISI 304 (1.4301). The outlet of the Stainless steel spigot turns 360° so it can be adjusted to any required position. With a practical suspension device for filling container and eyelet for shutting off the spigot.
Brand: Buerkle™ 0535-0020
Description
- Seals PTFE, FKM
- Self-closing
- Thread connection 3/4 in.
- Lockable
- NW 10 mm
Specifications
Silver | |
3/4 in. Thread Connection | |
Stainless steel |