Learn More
S Murray and Co™ Stainless Steel Spring Forceps
Use these forceps for applications such as major and minor surgery, post mortem, dissecting and dressing sets. S Murray™ Stainless Steel Spring Forceps comply with the requirements of the Medical Devices Directive 93/42/EEC, with full traceability maintained throughout the manufacturing process.
Brand: S Murray and Co™ E004/01
Description
Use these forceps for applications such as major and minor surgery, post mortem, dissecting and dressing sets. S Murray™ Stainless Steel Spring Forceps comply with the requirements of the Medical Devices Directive 93/42/EEC, with full traceability maintained throughout the manufacturing process.
- Stainless Steel forceps for dissection
Specifications
Spring Forceps | |
For dissection | |
Sharp, Narrow Flutes |
Stainless Steel | |
Each | |
115 mm |