missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Stainless Steel Tank, 12L

For water bath.

Brand:  Fisherbrand™ 17215556

Product Code. 17215556

  • 599.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

For use with Fisherbrand™ Digital Heating Circulator.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

12 L
410 x 410 x 500 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Stainless Steel Tank, 12L >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.