Learn More
RETSCH Stainless-Steel Test Sieve Stacks
Brand: RETSCH 60.166.000998
This item is not returnable.
View return policy
Specifications
1 Each | |
400 mm | |
Stainless Steel | |
Test Sieve Stack | |
Certificate of compliance with ISO 3310/1 or NFX 11-504, 10 Sieves (63μm, 125μm, 250μm, 500μm, 1mm, 2mm, 4mm, 8mm, 16mm, 31.5mm) and collecting pan |
For separation, fractioning, particle size determination | |
Stainless Steel | |
63μm, 125μm, 250μm, 500μm, 1mm, 2mm, 4mm, 8mm, 16mm, 31.5mm | |
ISO 3310 and ASTM E 11 | |
65mm |