Learn More
Spectrum™ Labs Standard RC Dialysis Disc
For standard grade RC dialysis tubing
Brand:  Spectrum™ Labs 132484
 View more versions of this product
Product Code. 12767526
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
For use with Spectrum Equilibrium Dialyzer and Spectra/Por™ MacroDialyzer systems.
  • Regenerated cellulose (RC) precut membrane discs
  • Supplied dry with a glycerol as a humectant
Specifications
Specifications
Standard RC Dialysis Disc
33 mm
50 Pk.
For standard grade RC dialysis tubing
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Spectrum™ Labs Standard RC Dialysis Disc > Diameter: 33mm