Learn More
IBA Solutions For Life Science StarGate™ Upstream Fusion Vector pNFUSE-IBA
For upstream positioning of an inserted gene in the final Fusion Donor vector when linking two or more genes cloned in separate Donor Vectors using the StarGate™ transfer cloning system.
213.00 €
Specifications
Format | Solution, liquid |
---|---|
For Use With (Equipment) | Stargate™ system derivatives |
Description | Resistance: ampicillin |
pH | 8 |
Product Type | Fusion Vector |
Product Code | Brand | Vector | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Vector | Price | Quantity & Availability | ||||||
15693718
|
IBA Solutions For Life Science
5-1631-001 |
pNFUSE-IBA w/ IRES11 | N/A | N/A | N/A | |||||
15603728
|
IBA Solutions For Life Science
5-1632-001 |
pNFUSE-IBA w/ SD11 | N/A | N/A | N/A | |||||
15613728
|
IBA Solutions For Life Science
5-1633-001 |
pNFUSE-IBA w/ LINK11 | N/A | N/A | N/A | |||||
15623728
|
IBA Solutions For Life Science
5-1634-001 |
pNFUSE-IBA w/ LINK12 | N/A | N/A | N/A | |||||
Description
StarGate™ offers a simple cloning procedure for rapid and highly efficient subcloning of an arbitrary gene of interest (GOI) for use in functional expression, purification, detection, and immobilization or separation of recombinant proteins.
- Minimal extra modification of the gene due to short combinatorial sites
- Inherent high-level cloning efficiency due to a directed reaction (no equilibrium)
- Genes are cloned in a Donor Vector and then parallel-transfered into Acceptor Vectors that provide different genetic surroundings
- Easily connect two or more genes that are cloned in separate Donor Vectors using Fusion Vectors that are assembled by use of a one-tube reaction
- The GOI to be positioned upstream is transferred into a pNFUSE-IBA derivative, specifying the intergenic region (IR):
- IRES11: Provides internal ribosomal entry site (IRES) from encephalomyocarditis virus for separate cap-independent translation of downstream cistron in mammalian cells
- SD11: IR includes a Shine Dalgarno (SD) sequence for translation of downstream cistron at comparable level to the upstream cistron in bacterial cells
- LINK11: Genes of interest are fused by a sequence encoding the 10 amino acid linker GlySerGlyGlyGlySerGlyGlyGlySer
- LINK12: Genes of interest are fused by a sequence encoding the 15 amino acid linker GlySerGlyGlySerGlyGlyGlyGlySerGlyGlyGlyGlySer
- The GOIs to be positioned downstream are transferred into a pCFUSE-IBA11 vector (identical, irrespective of IR)
Specifications
Solution, liquid | |
Resistance: ampicillin | |
Fusion Vector | |
2°C to 8°C for frequent usage, -20°C for long-term storage | |
5 μg | |
Upstream positioning, fusing cloned genes of interest |
Stargate™ system derivatives | |
8 | |
20μL TE buffer: 10mM Tris-HCl, 1mM EDTA per 5μL pCFUSE-IBA11 | |
250 ng/μL | |
Clear |
Safety and Handling
missing translation for 'shelfLife' : 12 months
Your input is important to us. Please complete this form to provide feedback related to the content on this product.