Learn More
Corning™ Sterile Polystyrene Inoculating Loops
GREENER_CHOICE
Constructed of flexible polystyrene for optimum retrieval of an inoculum from a culture. Gosselin™ Sterile Polystyrene Inoculating Loops provide a superior solution for transferring microscopic organisms, inoculum, or streaking.
Brand:  Corning™ OS1B-01
 View more versions of this product
Product Code. 15241065
  • 521.12 € / Pack of 9000
Estimated Shipment: 09-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Polystyrene (PS) inoculating loops
  • Spherical or needle extremity
  • 1 μl or 10 μl
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 μL
Polystyrene
White
3000/Pk.
195 mm
Sterile
Inoculating Loop, Spherical Tip
Tamper evident zip bag
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Corning™ Sterile Polystyrene Inoculating Loops > 1 μL; Spherical