Learn More
Sterilized Sample Ports for CTS™ Rotea™ Single-Use Kit
The Gibco CTS Rotea Single-Use Kit is part of the Gibco CTS Rotea Counterflow Centrifugation System. Each kit can perform a wide range of cell processing steps, making it ideal for wash & concentration, separation, isolation, and more.
Brand:  Invitrogen A50111
Product Code. 17206573
  • 405.00 € / Each
Explore more special offers
Description
Description
  • The Gibco CTS Rotea Single-Use Kit is part of the Gibco CTS Rotea Counterflow Centrifugation System
  • Each kit can perform a wide range of cell processing steps, making it ideal for wash & concentration, separation, isolation, and more
  • The single-use kit provides flexibility for process development and easily supports commercial-scale manufacturing
  • Each kit is compatible with sterile luer connectors or thermal welding
  • These Sterilized Sample Ports for use with the kit enable collection of product samples during protocol execution, for process development
  • Sample port assembly consists of a needleless port joined with short lengths of PVC tube
  • PVC tube is sealed to enable thermal welding
  • Sample port assemblies and PVC tubing are gamma-irradiated and individually packaged
TRUSTED_SUSTAINABILITY
Specifications
Specifications
10 Pk.
No
CTS Rotea
10 ports
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Sterilized Sample Ports for CTS™ Rotea™ Single-Use Kit > 10 Pk.