missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ SteriWare™ Powder Funnels
BIOPHARMA

These disposable SteriWare™ Powder Funnels are the ideal tools when loading powders, granules and pastes. The wide spout ensures effortless loading without costly bridging problems. They are available with full certification and documentation.

Brand:  Sampling Systems™ 8111P-350

Product Code. 17524606

  • 109.00 € / Pack of 8
Estimated Shipment: 06-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Disposable Powder Funnels are ideal for transferring powders and granules.
  • These powder funnels are supplied individually bagged and are available both sterile and non-sterile.
  • The funnels are made from chemically resistant, food grade HDPE and PP.
  • The wide opening at the base of the funnel will help prevent powder blockages.
  • The range is made from FDA, EC 1935/2004 and Food Grade EU 10/2011 conforming HDPE and PP.
  • The 200 mm size is moulded and packed in a Cleanroom for superior cleanliness.
  • Made from high quality PP.
  • Conforms to the latest FDA and EU food requirements.
  • Supplied individually bagged.
  • The 350 mm size is made outside the cleanroom and then taken into a cleanroom where its is checked and packed.
  • The funnels have a rigid handle.
  • Each batch is fully traceable.
Specifications

Specifications

350 mm
Polypropylene
Individually Bagged
40 mm
Non-sterile
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sampling Systems™ SteriWare™ Powder Funnels > SteriWare Powder Funnels; Diameter (Metric)Top: 350 mm; Diameter (Metric) Stem: 40 mm; Sterility: No; Material: Polypropylene

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.