missing translation for 'onlineSavingsMsg'
Learn More

Fistreem International™ Storage Reservoir

Replacement storage reservoir for the Cyclon Ultrapure Water Still. Fistreem International™ Storage Reservoir holds a full 30L distilled water.

Brand:  Fistreem International™ WSC900.RTB.9

Product Code. 13106020

  • 1205.00 € / Each
Estimated Shipment: 09-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Incorporates a fast flow tap and a sight tube showing the contents of the reservoir.
  • Distillate access ports are fitted on both sides, allowing the reservoir to be sited to either side of the still.
  • To ensure highest purity, the reservoir features a polyethylene liner, vent filter and a cleaning access port.
  • An electronic float switch shuts off the still on a full reservoir
Specifications

Specifications

750 mm
Storage Reservoir
300 mm
30 L
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fistreem International™ Storage Reservoir > Capacity: 30L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.