Learn More
Bryson Packaging™ Aluminum Foil Rolls
Ideal for use in autoclaves. Bryson Packaging™ Aluminum Foil Rolls are suitable for many laboratory applications including sealing bottles, culture tubes, stopping leaks, reducing evaporation or heat losses, covering corks and making weighing boats or dishes.
Brand: Bryson Packaging™ AKL-300-045W
Description
- Available in a variety of widths
- Can be cut to any length, cutter provided with box
- Sealing, weighing and transferring samples
Specifications
Ideal for use in autoclaves; 'Laboratory applications -including sealing bottles, culture tubes, stopping leaks, reducing evaporation or heat losses, covering corks and making weighing boats or dishes | |
600mm | |
12 μm |
Foil | |
Aluminum |
Product Suggestions
Customers who viewed this item also viewed