Learn More
Molecular Probes™ Streptavidin, lyophilized powder
A bacterially derived biotin-binding protein that reportedly exhibits less nonspecific binding than avidin.
Brand: Molecular Probes™ 434301
149.15 EUR valid until 2025-12-31
Use promo code "24369" to get your promotional price.
Description
- A bacterially derived biotin-binding protein that reportedly exhibits less nonspecific binding than avidin.
- Binding activity of streptavidin is determined to be >11 units per mg protein
- One unit of binding activity is defined as that amount of protein which binds 1 μg of biotin (Green, N.M., Biochem. J., 89:599,1963)
- Streptavidin is provided in lyophilized form in 10 mM phosphate buffered saline, pH 7.4
- Reconstitute with 1 mL of distilled or deionized water
- Store at 4°C

Specifications
Unconjugated | |
Streptavidin is provided in lyophilized form in 10 mM phosphate buffered saline, pH 7.4. Reconstitute with 1 ml of distilled or deionized water. Store at 4°C. | |
Lyophilized |
1 mg | |
Wet Ice | |
Streptavidin Conjugate (fluorescent) |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.