missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Streptavidin, lyophilized powder

A bacterially derived biotin-binding protein that reportedly exhibits less nonspecific binding than avidin.

164.00 € - 554.00 €

Specifications

Conjugate Unconjugated
Content And Storage Streptavidin is provided in lyophilized form in 10 mM phosphate buffered saline, pH 7.4. Reconstitute with 1 ml of distilled or deionized water. Store at 4°C.
Shipping Condition Wet Ice
Form Lyophilized
Product Type Streptavidin Conjugate (fluorescent)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10354073
View Documents
Molecular Probes™
434302
5 mg
554.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
10394023
View Documents
Molecular Probes™
434301
1 mg
164.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • A bacterially derived biotin-binding protein that reportedly exhibits less nonspecific binding than avidin.
  • Binding activity of streptavidin is determined to be >11 units per mg protein
  • One unit of binding activity is defined as that amount of protein which binds 1 μg of biotin (Green, N.M., Biochem. J., 89:599,1963)
  • Streptavidin is provided in lyophilized form in 10 mM phosphate buffered saline, pH 7.4
  • Reconstitute with 1 mL of distilled or deionized water
  • Store at 4°C
Specifications

Specifications

Unconjugated
Wet Ice
Streptavidin Conjugate (fluorescent)
Streptavidin is provided in lyophilized form in 10 mM phosphate buffered saline, pH 7.4. Reconstitute with 1 ml of distilled or deionized water. Store at 4°C.
Lyophilized
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Molecular Probes™ Streptavidin, lyophilized powder

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.