missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ Hotplate/Stirrer Retort Rod
Retort Rod for your hot plate, stirrer, and/or stirring hot plate.
Brand: Cole-Parmer™ 04805-91
This item is not returnable.
View return policy
Description
- SR1 Retort Rod
Specifications
Stuart™ UC152 and US152 Stirrer Hotplates | |
600 mm | |
Rod |
2 ft. | |
SR1 | |
Stainless Steel |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
CE Certificate
File size: 307.8 kB
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cole-Parmer™ Hotplate/Stirrer Retort Rod > Hotplate/Stirrer Retort Rod; For Use With: Stuart™ UC152 and US152 Stirrer Hotplates
Spot an opportunity for improvement?Share a Content Correction