missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sulfuric Acid Solution 0.5M (1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical
Click to view available options
Quantity:
6 Ampules
Packaging:
Ampoule
Specifications
Specifications
pH | 1 |
Color | Colorless |
CAS | 7664-93-9 |
Boiling Point | 120°C |
Packaging | Ampoule |
Quantity | 6 Ampules |
Molecular Formula | H2O4S |
Physical Form | Liquid |
MDL Number | MFCD00064589 |
Synonym | oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acide sulfurique, electrolyte acid, acidum sulfuricum, vitriol brown oil |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Documents
Scientific Resources
Brochures
Fisherbrand Focus on pH and electrochemistry
File size: 9.1 MB
Fisherbrand Focus on pH and electrochemistry
File size: 2.5 MB
Safety and Handling

Product Identifier
- Sulfuric acid, 5M solutrate concentrate for volumetric analysis (dilute 10x = 0.5M,(1N) working concentration)
- Danger
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- P234-Keep only in original container.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sulfuric Acid Solution 0.5M (1N), NIST Standard Concentrate, for Volumetric Analysis, Fisher Chemical >
Spot an opportunity for improvement?Share a Content Correction