Learn More
3M™ Easy Trap™ Sweep & Dust Sheets
Reduce the risk of bad impressions by trapping up to 8x more dirt, dust, and sand than traditional dust mops!
Brand:  3M™ Easy Trap™ 7100104366
Product Code. 17118710
  • 138.00 € / 1 roll
Estimated Shipment: 03-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Convenient, disposable sheets mean no laundering
  • Use under any flat floor tool or high duster
  • Sheets are two-sided and come in rolls perforated every 6 in.
  • Low drag reduces worker fatigue
  • Works in wet environments, and on most horizontal surfaces
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Duster Sheets
0.67 x 0.50 x 125 ft.
Trapping Dust, Dirt, Sand, and Hair
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
3M™ Easy Trap™ Sweep & Dust Sheets > Duster Sheets