Learn More
Fisherbrand™ SYBR™ Green qPCR Master Mix
Fisherbrand™ SYBR™ Green qPCR Master Mix is a 2X, ready-to-use master mix designed for dye-based, quantitative amplification of DNA (cDNA and gDNA) targets by real-time PCR.
Brand:  Fisherbrand™ A59530
 View more versions of this product
Product Code. 17806238
  • 2425.00 € / 50mL
Explore more special offers
Description
Show More
Specifications
Specifications
SYBR Green dye, DNA polymerase, with an antibody-mediated hot start, Heat-labile uracil-DNA glycosylase (UDG), ROX dye (passive reference dye), dNTP blend containing dUTP/dTTP, Optimized buffer components; Store at -20°C ± 10°C in the dark
Frozen
qPCR, 2-step RT-qPCR, RT-qPCR
SYBR Green, FAM
Dry Ice
2X
50 mL
SYBR
High
Fast or Standard
Master Mix
Gene Expression, DNA Quantitation, Microbial Detection, Real Time PCR (qPCR)
5000 x 20 μL Reactions
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Manuals
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2806438 Certificate TypeCertificate of Analysis Date16/01/2025 Catalog NumberA59530
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ SYBR™ Green qPCR Master Mix > Volume: 50 mL