missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Upright ULT Freezer Racks

Fisherbrand inventory control racks are ideal for all cold storage application. They fit all standard types of upright ULT freezers.

Brand:  Fisherbrand™ UF-542

 View more versions of this product

Product Code. 15709549

  • 123.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Holds standard cryoboxes
  • Made of high quality stainless steel
  • Side opening with handles on the front for easy box retrieval.
Specifications

Specifications

Side Access Freezer Rack
Stainless Steel
Upright ULT Freezer
20 (2 in.) boxes
Standard 2 in. Cryoboxes
679 x 140 x 225 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Upright ULT Freezer Racks > Holds: 20 (2 in.) boxes

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.