missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Tacta™ Mechanical Pipet Multipacks
BIOPHARMAGREENER_CHOICE

Multipacks combine a set of Tacta single-channel mechanical pipets with matching tip trays and useful accessories. Sartorius Tacta Mechanical Pipet Multipacks provides best alternative to replace your used mechanical pipets.

Brand:  Sartorius LH-729672

1129.99 EUR valid until 2025-12-31
Use promo code "24294" to get your promotional price.


 View more versions of this product

Product Code. 15643104

  • 1565.00 € / 1 set
Estimated Shipment: 27-05-2025
to see stock.
Explore more special offers
Add to basket

Includes: 4 x Single-channel pipets (0.5 to 10μL, 2 to 20μL, 20 to 200μL, 100 to 1000μL), Tip Racks of 96 tips (0.1 to 10μL, 0.5 to 200μL, 10 to 1000μL), 1 x Linear Stand, 4 x Single Pipet Holders
This item is not returnable. View return policy

Description

Description

  • Comfortable to hold due to ergonomically designed handle
  • Low pipetting and tip ejection forces
  • Smooth tip ejection with the new Optiject™ technology
  • Unique Optilock™ system provides flexibility for volume adjustment and locking
  • Calibration adjustment to provide accurate results for various liquid types
  • Reliable results, even over lengthy pipetting periods
  • Easy-to-read, four-digit volume display helps to set exact volumes
  • Easy to clean with only three parts to disassemble
  • Fully autoclavable
  • High-quality standard Optifit tips for various applications

Services

Two years

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Sartorius Tips
Reusable
1
Non-sterile
0.5 to 10 μL, 2 to 20 μL, 20 to 200 μL, 100 to 1000 μL
4 x Single-channel pipets (0.5 to 10μL, 2 to 20μL, 20 to 200μL, 100 to 1000μL), Tip Racks of 96 tips (0.1 to 10μL, 0.5 to 200μL, 10 to 1000μL), 1 x Linear Stand, 4 x Single Pipet Holders
High
138 mm (1-channel, 1000μL); 240 mm (8-channel, 300μL)
Autoclavable
Tacta
Tacta
Mechanical Pipette Multipacks
Four-digit volume display
10/Pk.
2 Years
79 g (1-channel, 1000μL); 138 g (8-channel, 300μL)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sartorius Tacta™ Mechanical Pipet Multipacks > Tacta 4-Pack; Pack of 10

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.