Learn More
Thermo Scientific™ Tango™ DRD1-bla U2OS Cells
The Tango™ DRD1-bla U2OS cells contain the human Dopamine Receptor 1 (DRD1) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line.
Brand: Thermo Scientific™ K1485
Description
The Tango™ DRD1-bla U2OS cells contain the human Dopamine Receptor 1 (DRD1) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line. This parental cell line stably expresses a beta-arrestin/TEV protease fusion protein and the beta-lactamase (bla) reporter gene under the control of a UAS response element.
The Tango™ DRD1-bla U2OS cell line has been functionally validated for Z'-factor and EC50 concentrations of dihydrexidine. The DRD1 cells have been tested for assay performance under variable assay conditions. These data are available in the Validation & Assay Performance Summary in the Manuals and Brochures section.

Specifications
NM_000794.2 | |
Dividing Cells | |
Tail from AVPR2 GPCR | |
GPCRs | |
DRD1-bla U2OS cells, DRD1-bla U2OS Assay Protocol, Certificate of Analysis | |
Tango™ | |
BLA (Beta-Lactamase) | |
U2OS Cell | |
Tango™ |
Cell-based beta-arrestin recruitment | |
LiveBLAzer™-FRET B/G Loading Kit, Solution D and other materials are required separately; please refer to the protocol. Includes: • Tango™ DRD1-bla U2OS cells • Tango™ DRD1-bla U2OS Assay Protocol • Certificate of Analysis The Tango™ DRD1-bla U2OS cells are shipped on dry ice and should be stored in liquid nitrogen immediately upon receipt. |
|
Fluorescence | |
DRD1 | |
Single Cell Clone Origin | |
2 x 106 Cells | |
DRD1, D1 | |
U2OS | |
GPCR |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
40677 | Certificate of Analysis | 16/04/2017 | K1485 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.