missing translation for 'onlineSavingsMsg'
Learn More

TaqMan™ TaqMan™ Genotyping Master Mix

Optimized for end-point fluorescence detection in SNP genotyping applications.

Brand:  TaqMan™ 4381657

 View more versions of this product

Product Code. 10239534

  • 9120.00 € / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Distinct clusters and high call rates for unambiguous allelic discrimination
  • Validated with TaqMan™ SNP Genotyping Assays
  • Excellent pre- and post-PCR stability for high throughput setup and analysis
  • Reaction size is based on a 50μL reaction volume

May be used for a variety of genotyping applications, including:
  • Candidate gene studies
  • Drug-target validation
  • Disease association studies
  • Population genetics
  • Linkage mapping
  • Agricultural applications

Exceptional Performance from an Optimized Mix (2X)
Accurate genotype assignments result from preferential binding of the allele-specific probe to the matching target. TaqMan™ Genotyping Master Mix enables specific binding of the probe to achieve exceptional cluster resolution. Key features include:
  • AmpliTaq Gold™ DNA Polymerase, UP (Ultra Pure) - Automatic hot start enzyme designed to be active during thermal cycling and inactive at room temperature for easy reaction setup
  • Optimized mix components provide excellent specificity for discrimination between alleles
  • Passive internal reference based on proprietary ROX™ dye for increased precision on Applied Biosystems™ real-time PCR instruments
  • Single thermal cycling condition for consistent results with TaqMan™ Assays

Validated with TaqMan™ Genotyping Assays and Instruments:
  • TaqMan™ Genotyping Master Mix has been tested across all types of TaqMan™ SNP Genotyping Assays: TaqMan™ Drug Metabolism Genotyping Assays, TaqMan™ SNP Genotyping Assays, and Custom TaqMan™ SNP Genotyping Assays
  • In addition, the master mix is validated with the Applied Biosystems™ thermal cyclers and real-time PCR systems

High-Throughput Setup and Analysis:
  • The combination of several low-cost thermal cyclers for PCR and a single real-time PCR instrument for allelic discrimination make high throughput SNP genotyping manageable
  • TaqMan™ Genotyping Master Mix further improves high-throughput setup and analysis with excellent room temperature stability both before and after PCR
  • This provides the flexibility required to run long experiments, unattended overnight or over a weekend

Reliable Discrimination with Challenging Targets
  • Amplicon design constraints make SNP detection challenging, particularly when targets contain an abundance of GC- or AT-rich regions
  • The optimized components of TaqMan™ Genotyping Master Mix result in tight, well-separated clusters and more accurate allele-calls compared to other mixes, even for challenging targets

Setting the Industry Standard for Quality:
  • Extensive analytical and functional tests are performed for each manufactured lot of TaqMan™ Genotyping Master Mix, including one TaqMan™ SNP Genotyping Assay and one TaqMan™ Drug Metabolism Genotyping Assay
  • Results are compiled in an informative Certificate of Analysis, which is accessible through the Applied Biosystems™ Web site

Order Info

Guaranteed minimum shelf life is 60 days. Shipping Conditions: Wet Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Supplied at 2X concentration. The mix is optimized for SNP genotyping using TaqMan™ probes and contains AmpliTaq Gold™ DNA Polymerase UP (Ultra Pure), dNTPs without dUTP, Passive Reference 1 and optimized mix components. This pack contains two 50 mL bottles, sufficient for 4,000 reactions at the 50 μL total volume.

Storage: 2-8°C

Guaranteed minimum shelf life is 60 days (exact expiry date printed on product and CofA).
High
AmpliTaq Gold DNA Polymerase
SNP Genotyping (Real-Time PCR-Based), CNV (Copy Number Variation)
Copy Number Variants, SNPs (Unknown or Numerous), Drug Metabolizing Enzymes, SNPs (Known)
TaqMan™
UP (Ultra Pure)
Genotyping
2 X
100 mL
Primer-probe
qPCR
Standard
Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, QuantStudio™ 12k Flex, QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Flex, QuantStudio™ 7
ROX (Pre-mixed)
Genotyping Master Mix
2 x 50 mL
2X
DNA (Genomic)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.