Learn More
TFA Dostmann Temperature/Humidity Transmitter
Transmitter for TFA wireless thermo-hygrometers and weather stations.
Brand:  TFA Dostmann 30.3187.IT
Product Code. 17754239
  • 21.95 € / Each
Estimated Shipment: 03-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Transmitter for TFA wireless thermo-hygrometers and weather stations: 30.3044, 30.3045, 35.1109, 35.1111, 35.1113, 35.1121
  • For wireless transmission of the outdoor climate
  • Display with temperature and humidity
  • Range: 100 m in open field
  • Update: every 4 seconds
  • Also for climate monitoring of remote rooms
Specifications
Specifications
Temperature/Humidity Transmitter
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
TFA Dostmann Temperature/Humidity Transmitter > Temperature/Humidity Transmitter