missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl chloride, 98+%, Thermo Scientific Chemicals

27.84 € - 55.17 €

Chemical Identifiers

CAS 507-20-0
Molecular Formula C4H9Cl
Molecular Weight (g/mol) 92.566
MDL Number MFCD00000816
InChI Key NBRKLOOSMBRFMH-UHFFFAOYSA-N
Synonym tert-butyl chloride, propane, 2-chloro-2-methyl, tert-butylchloride, chlorotrimethylmethane, trimethylchloromethane, 2-chloroisobutane, t-butyl chloride, t-butylchloride, 2-methyl-2-chloropropane, 2-methyl-2-propyl chloride
PubChem CID 10486
IUPAC Name 2-chloro-2-methylpropane
SMILES CC(C)(C)Cl
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11469903
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A13004.AK
250 mL
27.84 €
250mL
Please to purchase this item. Need a web account? Register with us today!
11309659
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A13004.AP
500 mL
33.47 €
500mL
Please to purchase this item. Need a web account? Register with us today!
11459903
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A13004.AU
1000 mL
55.17 €
1000mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

tert-Butyl chloride plays an important role as a starting material to perform nucleophilic substitution reactions in order to prepare alcohol and alkoxides salts. It is used as an alkylating agent for the introduction of tert-butyl group and is also involved in Friedel-Crafts reactions. It is employed as an intermediate for the synthesis of agrochemicals and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl chloride plays an important role as a starting material to perform nucleophilic substitution reactions in order to prepare alcohol and alkoxides salts. It is used as an alkylating agent for the introduction of tert-butyl group and is also involved in Friedel-Crafts reactions. It is employed as an intermediate for the synthesis of agrochemicals and pharmaceuticals.

Solubility
Miscible with alcohol, benzene, chloroform and ether. Slightly miscible with water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong bases.
Specifications

Chemical Identifiers

507-20-0
92.566
NBRKLOOSMBRFMH-UHFFFAOYSA-N
10486
CC(C)(C)Cl
C4H9Cl
MFCD00000816
tert-butyl chloride, propane, 2-chloro-2-methyl, tert-butylchloride, chlorotrimethylmethane, trimethylchloromethane, 2-chloroisobutane, t-butyl chloride, t-butylchloride, 2-methyl-2-chloropropane, 2-methyl-2-propyl chloride
2-chloro-2-methylpropane

Specifications

507-20-0
0.846
−27°C (−16°F)
C4H9Cl
MFCD00000816
1730872
tert-butyl chloride, propane, 2-chloro-2-methyl, tert-butylchloride, chlorotrimethylmethane, trimethylchloromethane, 2-chloroisobutane, t-butyl chloride, t-butylchloride, 2-methyl-2-chloropropane, 2-methyl-2-propyl chloride
NBRKLOOSMBRFMH-UHFFFAOYSA-N
2-chloro-2-methylpropane
10486
≥98%
-26°C
50°C to 52°C
Mild
1.3848
UN1127
14,1562
Miscible with alcohol,benzene,chloroform and ether. Slightly miscible with water.
CC(C)(C)Cl
92.566
92.57
tert-Butyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225
Highly flammable liquid and vapour.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H225

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: CHLOROBUTANES

missing translation for 'einecsNumber' : 208-066-4

missing translation for 'rtecsNumber' : TX5040000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • tert-Butyl chloride

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

Supplemental information
  • MIXTURE LIST-Contains : tert-Butyl chloride

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only