missing translation for 'onlineSavingsMsg'
Learn More
Learn More
tert-Butyl N-(3-formylbenzyl)carbamate, 90%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
Assay Percent Range | 90% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Synonym | tert-butyl 3-formylbenzylcarbamate, tert-butyl n-3-formylphenyl methyl carbamate, tert-butyl 3-formylbenzyl carbamate, 3-aminomethyl benzaldehyde, n-boc protected, 3-formylbenzylamine, n-boc protected, tert-butyl n-3 formylbenzyl carbamate, tert-butyl n-3-formylbenzyl carbamate, tert-butyl3-formylbenzylcarbamate, 3-formylbenzyl carbamic acid tert-butyl ester, 3-formyl-benzyl-carbamic acid tert-butyl ester |
Formula Weight | 235.28 |
Percent Purity | 90% |
Chemical Name or Material | tert-Butyl 3-formylbenzylcarbamate |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
tert-Butyl N-(3-formylbenzyl)carbamate, 90%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction