Learn More
Product Code | Brand | Capacity (Metric) | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Length (Metric) | Diameter (Metric) | Price | Quantity & Availability | ||||
17981142
|
Fisherbrand™
90106188 |
5 mL | 75 mm | 12 mm |
24.50 €
Pack of 100 |
In Stock
Log in to see stock. ![]() |
||||
17901142
|
Fisherbrand™
90106179 |
12 mL | 130 mm | 14 mm |
36.40 €
Pack of 100 |
In Stock
Log in to see stock. ![]() |
||||
17991122
|
Fisherbrand™
90106189 |
13 mL | 100 mm | 16 mm |
40.50 €
Pack of 100 |
Estimated Shipment: 28-04-2025
Log in to see stock. |
||||
15822295
|
Fisherbrand™
90106160 |
17 mL | 125 mm | 16 mm |
46.85 €
Pack of 100 |
Estimated Shipment: 22-05-2025
Log in to see stock. |
||||
17951122
|
Fisherbrand™
90106180 |
17 mL | 130 mm | 16 mm |
51.25 €
Pack of 100 |
In Stock
Log in to see stock. ![]() |
||||
15802285
|
Fisherbrand™
90106133 |
21 mL | 150 mm | 16 mm |
62.60 €
Pack of 100 |
In Stock
Log in to see stock. |
||||
17931122
|
Fisherbrand™
90106181 |
21 mL | 160 mm | 16 mm |
60.50 €
Pack of 100 |
In Stock
Log in to see stock. ![]() |
||||
17961132
|
Fisherbrand™
90106190 |
27 mL | 150 mm | 18 mm |
63.54 €
Pack of 100 |
In Stock
Log in to see stock. |
||||
17921142
|
Fisherbrand™
90106182 |
32 mL | 180 mm | 18 mm |
65.05 €
Pack of 100 |
In Stock
Log in to see stock. ![]() |
||||
17941122
|
Fisherbrand™
90106183 |
34 mL | 150 mm | 20 mm |
64.60 €
Pack of 100 |
In Stock
Log in to see stock. ![]() |
||||
17941152
|
Fisherbrand™
90106191 |
53 mL | 150 mm | 24 mm |
35.90 €
Pack of 50 |
Estimated Shipment: 28-04-2025
Log in to see stock. |
||||
17931142
|
Fisherbrand™
90106184 |
57 mL | 150 mm | 25 mm |
37.95 €
Pack of 50 |
Estimated Shipment: 28-04-2025
Log in to see stock. |
||||
17911142
|
Fisherbrand™
90106185 |
112 mL | 200 mm | 30 mm |
46.55 €
Pack of 50 |
In Stock
Log in to see stock. ![]() |
||||
Description
- Transferring substances and liquids into other states of aggregation and thus initiating further reactions is very important for the daily work in the laboratory. A wide variety of vessels, apparatus and equipment are used for these work. Reaction controls, test series and preliminary samples are usually carried out in test tubes due to the small amounts of substance removed.
- Manufactured from borosilicate glass 3.3
- Thick-walled and therefore mechanically very resistant
- With round bottom
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Fisherbrand™ Test Tubes, Borosilicate Glass 3.3, Without Rim, Medium Wall