missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran, BAKER ANALYZED, J.T. Baker™

High quality grade chemical stabilized with 0.025% BHT for laboratory and specialized industrial use.

90.00 € - 1185.00 €

Chemical Identifiers

CAS 109-99-9
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00005356
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
Synonym tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
PubChem CID 8028
ChEBI CHEBI:26911
IUPAC Name oxolane
SMILES C1CCOC1
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10425191
View Documents
JT Baker
8075.1000
1 L Glass
90.00 €
1 L
Please to purchase this item. Need a web account? Register with us today!
10455381
View Documents
JT Baker
8075.5000
5 L EcoTainer Metal Safety Can This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10465571
View Documents
JT Baker
8075.9025
25 L Steel
1185.00 €
25 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Label provides an actual Lot Analysis as a guarantee of purity and consistency
  • Analytical methods used for these determinations are based on standard test procedures described in technical publications
  • Improvements developed by J.T. Baker are employed to achieve a higher degree of reliability
Specifications

Chemical Identifiers

109-99-9
72.11
WYURNTSHIVDZCO-UHFFFAOYSA-N
8028
oxolane
C4H8O
MFCD00005356
tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
CHEBI:26911
C1CCOC1

Specifications

-108.5°C
Colorless
65°C to 67°C
−21°C
Liquid
C4H8O
MFCD00005356
tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
WYURNTSHIVDZCO-UHFFFAOYSA-N
oxolane
8028
72.11g/mol
BAKER ANALYZED™
109-99-9
0.880 to 0.883 g/mL at 25°C
1.5 - 12 Vol %
230°C
Max. 0.002%
OCH2CH2CH2CH2
Max. 0.02%
Fully miscible in water
C1CCOC1
72.11
CHEBI:26911
≥99%
Tetrahydrofuran
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 203-726-8

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • EUH019-May form explosive peroxides.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.