Learn More
Thermo Scientific™ H-6000A Bucket Adapters
Accept a variety of blood processing tubes, bags and bottles
Brand:  Thermo Scientific™ 01092
 View more versions of this product
Product Code. 15129485
  • 232.16 € / Each
Explore more special offers
Description
Description
Accept a variety of blood processing tubes, bags and bottles
For use with Thermo Scientific H-6000A buckets.
  • Adapters hold a maximum capacity of 6 × 500mL (bottles), 6 × 550mL (blood bags), and 132 × 7mL (tubes). Max. O.D. × L: 70 x 160mm
TRUSTED_SUSTAINABILITY
Specifications
Specifications
6 x 250 mL Bottles
H-6000A bucket for Thermo Scientific™ Sorvall™ RC3BP™ Plus low speed centrifuge rotor
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ H-6000A Bucket Adapters > 250mL bottle