missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ COD (Chemical Oxygen Demand) Calibration Standards for AQUAfast Colorimeters

Use these calibration standards to check your colorimeter instrument operations before measuring COD chemistries, for example in environmental and wastewater monitoring.

Brand:  Thermo Scientific™ CODS10

 View more versions of this product

Product Code. 11707757

  • 112.56 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Use Thermo Scientific™ COD (Chemical Oxygen Demand) Calibration Standards with Orion™ AQUAfast Colorimeters such as AQ4000 Advanced Colorimeter and Orion AQUAfast II AQ2040 Colorimeter. Orion colorimeters, which measure COD chemistries, are ideal for environmental and wastewater monitoring. Use these calibration standards to check operations prior to using the instruments.

  • For use with: Orion AQUAfast AQ4000, AQ3140 and AQ2040 colorimeters
  • Two Calibration Standards are available: 1000 ppm (CODS01) and 10000 ppm (CODS10), both in 475 mL bottles
  • Ideal for environmental and wastewater monitoring

Order Info

Reagents sold separately.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

475 mL
Environmental and Wastewater Monitoring
AQUAfast Colorimeter
EPA
10000 ppm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberBP1 Certificate TypeCertificate of Analysis Date22/11/2023 Catalog NumberCODS10
Lot NumberAV1 Certificate TypeCertificate of Analysis Date26/05/2022 Catalog NumberCODS10
Lot NumberZP1 Certificate TypeCertificate of Analysis Date21/12/2021 Catalog NumberCODS10
Lot NumberYV1 Certificate TypeCertificate of Analysis Date19/05/2020 Catalog NumberCODS10
Lot NumberXQ1 Certificate TypeCertificate of Analysis Date10/10/2019 Catalog NumberCODS10
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ COD (Chemical Oxygen Demand) Calibration Standards for AQUAfast Colorimeters > Cod Standard 10000 Ppm 475ml

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.