missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Phusion Hot Start II High-Fidelity PCR Master Mixes

Thermo Scientific Phusion High-Fidelity DNA polymerases set a gold standard for high performance PCR.

219.00 € - 872.00 €

Specifications

GC-Rich PCR Performance Medium
Polymerase Phusion Hot Start II High-Fidelity DNA Polymerase
Reaction Speed Fast
Product Type Hot Start High-Fidelity PCR Master Mix
Shipping Condition Dry Ice
View More Specs

Products 4
Product Code Brand Color No. of Reactions Price Quantity & Availability  
Product Code Brand Color No. of Reactions Price Quantity & Availability  
15331732
View Documents
Thermo Scientific™
F566S
Green 100 Reactions
219.00 €
100 reactions
Please to purchase this item. Need a web account? Register with us today!
15341732
View Documents
Thermo Scientific™
F566L
Green 500 Reactions
862.00 €
500 reactions
Please to purchase this item. Need a web account? Register with us today!
15311732
View Documents
Thermo Scientific™
F565L
Colorless 500 Reactions
872.00 €
500 reactions
Please to purchase this item. Need a web account? Register with us today!
15301732
View Documents
Thermo Scientific™
F565S
Colorless 100 Reactions
221.00 €
100 reactions
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Thermo Scientific Phusion High-Fidelity DNA Polymerases set a gold standard for high performance PCR. Featuring an error rate 50-fold lower than that of Taq, and 6-fold lower than that of Pfu, Phusion High-Fidelity DNA Polymerase is excellent choice for cloning and other applications requiring high fidelity. Phusion DNA Polymerases offer robust performance with short protocol times, even in the presence of PCR inhibitors, and generate higher yields with lower enzyme amounts than other DNA polymerase.

Highlights

  • Reaction set up at room temperature
  • No non-specific amplification and primer degradation during reaction set up
  • Zero-time reactivation due to unique hot start technology
  • High fidelity (52X Taq)
  • Fast PCR due to short extension times (15-30 s/kb)
  • Robust reactions, minimal optimization needed
  • Increased product yields with minimal enzyme amounts
  • Direct loading on gels

With Phusion Hot Start II High-Fidelity DNA Polymerases amplification proceeds without the production of nonspecific products due to the combination of Phusion DNA Polymerase and a reversibly bound, specific Affibody ligand that inhibits DNA polymerase activity at room temperature. The Affibody ligand also inhibits the 3'→5' exonuclease activity of the polymerase, thus preventing degradation of primers and template DNA during reaction set up. At temperatures that promote polymerase activity, the ligand is released, rendering the polymerase fully active. Phusion Hot Start II DNA Polymerase is immediately reactivated at high temperatures so it does not require a separate activation step in PCR protocols.

Phusion Green Hot Start II High-Fidelity PCR Master Mix is convenient 2X mix designed to minimize the number of pipetting steps. The master mix contains Phusion Hot Start II DNA Polymerase, nucleotides and optimized reaction buffer including MgCl2. The buffer also includes a density reagent and two tracking dyes for direct loading of PCR products on a gel.

Applications

  • High-fidelity PCR
  • High throughput
  • Amplification of difficult (GC-rich) templates
  • Template generation for sequencing
  • Multiplex PCR
  • Long-range PCR
  • Cloning
  • Mutagenesis
  • Microarray

Using Phusion DNA Polymerases

Annealing rules for Phusion DNA Polymerases are different from many common DNA polymerases (such as Taq DNA polymerases).

Specifications

Specifications

Medium
Fast
Dry Ice
2X
Built-In Hot Start
SuperMix or Master Mix
Phusion Hot Start II High-Fidelity DNA Polymerase
Hot Start High-Fidelity PCR Master Mix
Hot-start PCR, High-fidelity PCR
52X
Blunt
20 kb or less
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.