missing translation for 'onlineSavingsMsg'
Learn More

Sonoco™ ThermoSafe PolarPack™ Standard

PolarPack refrigerants provide thermal solutions for temperature critical packages during transit. Available in a broad selection of styles and a multitude of sizes, our refrigerants are designed to provide reliable temperature protection for a vast array of temperature-sensitive products and packages.

Brand:  Sonoco™ PP16

 View more versions of this product

Product Code. 13099332

  • 39.00 € / Pack of 36
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Thermal solutions for temperature critical packages during transit
  • Gel will phase change at 0° C.
  • condition coolant in freezer before shipments.
  • refrigeration for transport of food, pharmaceuticals and medical products. 
  • For frozen or refrigerated shipments.
  • Alternative to wet ice.
  • Available in several sizes
TRUSTED_SUSTAINABILITY
Specifications

Specifications

16 oz.
5.5 in.
16 oz.
Polyethylene Film
For shipping temperature critical packages during transit
6.5 in.
1 in.
6.5 x 5.5 x 1 in.
0°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Sonoco™ ThermoSafe PolarPack™ Standard > 16 oz

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.