missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Thermostatted Single Cell Holder for GENESYS 140/150/180 & BioMate 160 Spectrophotometers
Control sampling temperatures for life science applications such as biochemical kinetics, enzymatic food analysis and equilibrium studies.
Brand: Thermo Scientific™ 222-238800
This item is not returnable.
View return policy
Description
Ideal for life science applications such as biochemical kinetics, enzymatic food analysis and equilibrium studies.
Peltier controlled sealed liquid system provides precise temperature control
little to no maintenance needed
150ml liquid volume means less mess from accidental spills

Specifications
TPS 1500W Sealed Peltier Recirculator | |
Control temperature with a water thermostatted cell holder. | |
± 0.05 °C | |
150 mL |
Sealed Peltier Recirculator System | |
GENESYS 140/150/180, BioMate 160 Spectrophotometers | |
20°C to 60 °C |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ Thermostatted Single Cell Holder for GENESYS 140/150/180 & BioMate 160 Spectrophotometers > Sealed Peltier Reciculator
Spot an opportunity for improvement?Share a Content Correction