missing translation for 'onlineSavingsMsg'
Learn More
Thieno[2,3-b]pyridin-2-ylmethanol, 97%, Thermo Scientific™ Product Code.: 10597663

Thieno[2,3-b]pyridin-2-ylmethanol, 97%, Thermo Scientific™

Product Code. 10597663
1 g, Amber Glass Bottle
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10597663

Brand: Thermo Scientific Maybridge CC05609DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 131337-81-0
Molecular Formula C8H7NOS
Molecular Weight (g/mol) 165.21
MDL Number MFCD07772792
InChI Key XLUKSWZEEAGBDH-UHFFFAOYSA-N
Synonym thieno 2,3-b pyridin-2-ylmethanol, thieno 2,3-b pyridine-2-methanol, thieno 2,3-b pyridin-2-yl methanol, acmc-1bz5x, thieno 2,3-b pyridine-2-ylmethanol, 2-hydroxymethylthieno 2,3-b pyridine, thiopheno 2,3-b pyridin-2-ylmethan-1-olShow MoreShow Less
PubChem CID 7162092
IUPAC Name thieno[2,3-b]pyridin-2-ylmethanol
SMILES C1=CC2=C(N=C1)SC(=C2)CO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 165.22
Percent Purity 97%
Chemical Name or Material Thieno[2, 3-b]pyridin-2-ylmethanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thieno[2,3-b]pyridin-2-ylmethanol, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.