missing translation for 'onlineSavingsMsg'
Learn More
Thieno[2,3-b]pyridine-2-carbaldehyde, ≥97%, Thermo Scientific™ Product Code.: 10750534

Thieno[2,3-b]pyridine-2-carbaldehyde, ≥97%, Thermo Scientific™

Product Code. 10750534
1 g, Amber Glass Bottle
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10750534

Brand: Thermo Scientific Maybridge CC05604DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 53174-98-4
Molecular Formula C8H5NOS
Molecular Weight (g/mol) 163.194
MDL Number MFCD07772791
InChI Key HGZQKYOLRLVMHZ-UHFFFAOYSA-N
Synonym thieno 2,3-b pyridine-2-carbaldehyde, thieno 2,3-b pyridine-2-carboxaldehyde, 2-formylthieno 2,3-b pyridine, thiopheno 2,3-b pyridine-2-carbaldehydeShow MoreShow Less
PubChem CID 289925
IUPAC Name thieno[2,3-b]pyridine-2-carbaldehyde
SMILES C1=CC2=C(N=C1)SC(=C2)C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 163.2
Percent Purity 97%
Chemical Name or Material Thieno[2, 3-b]pyridine-2-carbaldehyde
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thieno[2,3-b]pyridine-2-carbaldehyde, ≥97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.