missing translation for 'onlineSavingsMsg'
Learn More
Thiophenol, 99+%, Thermo Scientific Chemicals Product Code.: 11441970

Thiophenol, 99+%, Thermo Scientific Chemicals

Product Code. 11441970
250 g
Click to view available options
Quantity:
250 g
1000 g
This item is not returnable. View return policy

Product Code. 11441970

Brand: Thermo Scientific Alfa Aesar A15916.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thiophenol acts as a reagent to synthesize alfa, beta-unsaturated sulfides, beta-lactones and (3H-benzo[c][1,2]diselenol-3-yl)(phenyl)sulfane. It is used as an antinematodal agent and an intermediate in the preparation of sodium thiophenolate, pentachlorothiophenol and alkyl phenylsulfides. It acts as an inhibitor of human fibroblast collagenase in vitro.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiophenol acts as a reagent to synthesize alfa, beta-unsaturated sulfides, beta-lactones and (3H-benzo[c][1,2]diselenol-3-yl)(phenyl)sulfane. It is used as an antinematodal agent and an intermediate in the preparation of sodium thiophenolate, pentachlorothiophenol and alkyl phenylsulfides. It acts as an inhibitor of human fibroblast collagenase in vitro.

Solubility
Miscible with most organic solvents and aqueous base. Immiscible with water.

Notes
Incompatible with strong acids, bases and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-98-5
Molecular Formula C6H6S
Molecular Weight (g/mol) 110.174
MDL Number MFCD00004826
InChI Key RMVRSNDYEFQCLF-UHFFFAOYSA-N
Synonym thiophenol, phenyl mercaptan, mercaptobenzene, phenylthiol, phenylmercaptan, thiofenol, phenol, thio, thiophenate, thiofenol czech, benzene, mercaptoShow MoreShow Less
PubChem CID 7969
ChEBI CHEBI:48498
IUPAC Name benzenethiol
SMILES C1=CC=C(C=C1)S

Specifications

Melting Point °C to 15°C
Density 1.075
Boiling Point 168°C to 169°C
Flash Point 55°C (131°F)
Assay Percent Range ≥99%
Refractive Index 1.59
Quantity 250 g
UN Number UN2337
Beilstein 506523
Sensitivity Air Sensitive
Merck Index 14,9355
Solubility Information Miscible with most organic solvents and aqueous base. Immiscible with water.
Formula Weight 110.18
Percent Purity ≥99%
Physical Form Liquid
Chemical Name or Material Thiophenol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0431068 Certificate TypeCertificate of Analysis Date13/10/2022 Catalog NumberA15916.0B,A15916.30,A15916.18
Lot Number10243525 Certificate TypeCertificate of Analysis Date15/09/2022 Catalog NumberA15916.0B,A15916.30,A15916.18
Lot Number10219809 Certificate TypeCertificate of Analysis Date08/07/2022 Catalog NumberA15916.0B,A15916.30,A15916.18
Lot Number60900426 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15916.0B,A15916.30,A15916.18
Lot NumberG29N21 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA15916.0B,A15916.30,A15916.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thiophenol, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.