Learn More
Thiophenol, 99+%, Thermo Scientific Chemicals
Description
Thiophenol acts as a reagent to synthesize alfa, beta-unsaturated sulfides, beta-lactones and (3H-benzo[c][1,2]diselenol-3-yl)(phenyl)sulfane. It is used as an antinematodal agent and an intermediate in the preparation of sodium thiophenolate, pentachlorothiophenol and alkyl phenylsulfides. It acts as an inhibitor of human fibroblast collagenase in vitro.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Thiophenol acts as a reagent to synthesize alfa, beta-unsaturated sulfides, beta-lactones and (3H-benzo[c][1,2]diselenol-3-yl)(phenyl)sulfane. It is used as an antinematodal agent and an intermediate in the preparation of sodium thiophenolate, pentachlorothiophenol and alkyl phenylsulfides. It acts as an inhibitor of human fibroblast collagenase in vitro.
Solubility
Miscible with most organic solvents and aqueous base. Immiscible with water.
Notes
Incompatible with strong acids, bases and oxidizing agents.

Specifications
Specifications
Melting Point | -15°C |
Density | 1.075 |
pH | 5 |
Boiling Point | 168°C to 169°C |
Flash Point | 55°C (131°F) |
Assay Percent Range | ≥99% |
Odor | Stench |
Refractive Index | 1.59 |
Quantity | 1000 g |
UN Number | UN2337 |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0431068 | Certificate of Analysis | 13/10/2022 | A15916.0B,A15916.30,A15916.18 |
10243525 | Certificate of Analysis | 15/09/2022 | A15916.0B,A15916.30,A15916.18 |
10219809 | Certificate of Analysis | 08/07/2022 | A15916.0B,A15916.30,A15916.18 |
60900426 | Certificate of Analysis | 23/11/2021 | A15916.0B,A15916.30,A15916.18 |
G29N21 | Certificate of Analysis | 23/11/2021 | A15916.0B,A15916.30,A15916.18 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.