missing translation for 'onlineSavingsMsg'
Learn More
Tin(II) phthalocyanine, Thermo Scientific Chemicals Product Code.: 11384537

Tin(II) phthalocyanine, Thermo Scientific Chemicals

Product Code. 11384537
0.5 g
Click to view available options
Quantity:
0.5 g
2 g
This item is not returnable. View return policy

Product Code. 11384537

Brand: Thermo Scientific Alfa Aesar 039607.02

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Tin(II) phthalocyanine is used as an organic light-emitting diode (OLED) material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15304-57-1
Molecular Formula C32H16N8Sn
Molecular Weight (g/mol) 631.25
MDL Number MFCD00058867
InChI Key LLVONELOQJAYBZ-UHFFFAOYSA-N
Synonym tin ii phthalocyanine, tin phthalocyanine, snpc, phthalocyanatotin cento

Specifications

Quantity 0.5 g
UN Number UN3146
Solubility Information Insoluble in water.
Formula Weight 631.43
Physical Form Powder
Chemical Name or Material Tin(II) phthalocyanine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ02K056 Certificate TypeCertificate of Analysis Date04/12/2024 Catalog Number039607.02
Lot NumberY26K017 Certificate TypeCertificate of Analysis Date27/11/2024 Catalog Number039607.02
Lot NumberP27K033 Certificate TypeCertificate of Analysis Date04/04/2024 Catalog Number039607.02
Lot NumberP23J018 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog Number039607.02
Lot NumberU02D024 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number039607.02

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tin(II) phthalocyanine, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.