Learn More
Thermo Scientific™ TR-FRET Coregulator Buffer H
A specialized nuclear receptor buffer that can be used with Invitrogen's collection of purified nuclear receptor proteins and assays.
Brand: Thermo Scientific™ PV4661
Description
A specialized nuclear receptor buffer that can be used with Invitrogen's collection of purified nuclear receptor proteins and assays.

Specifications
TR-FRET Coregulator Buffer H, 25mL, Liquid, Store at 20 to 30°C, Water Soluble, Used For Pharma & Biopharma, Target & Lead Identification & Validation | |
25mL, 25 mL | |
TR-FRET Coregulator Buffer H may be shipped frozen or at room temperature. Store at room temperature (20°C to 30°C). |
Liquid | |
LanthaScreen™ Nuclear Receptor Cofactor Interaction Assays | |
Approved for shipment at Room Temperature or on Wet or Dry Ice |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2944408B | Certificate of Analysis | 21/02/2025 | PV4661 |
2944408C | Certificate of Analysis | 21/02/2025 | PV4661 |
2944408A | Certificate of Analysis | 21/02/2025 | PV4661 |
2561688D | Certificate of Analysis | 23/02/2024 | PV4661 |
2561688C | Certificate of Analysis | 22/08/2023 | PV4661 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.