missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TR-FRET Coregulator Buffer I

A specialized nuclear receptor buffer that can be used with Invitrogen's collection of purified nuclear receptor proteins and assays.

Brand:  Thermo Scientific™ PV4662

Product Code. 10122428

  • 154.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

A specialized nuclear receptor buffer that can be used with Invitrogen's collection of purified nuclear receptor proteins and assays.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

TR-FRET Coregulator Buffer I, 25mL, Liquid, 7.5 pH Range, Store at 20 to 30°C, Proprietary Buffer, Water Soluble, Used For Pharma & Biopharma, Target & Lead Identification & Validation
25mL, 25 mL
TR-FRET Coregulator Buffer I may be shipped frozen or at room temperature. Store at room temperature (20°C to 30°C).
Liquid
LanthaScreen™ Nuclear Receptor Cofactor Interaction Assays
Approved for shipment at Room Temperature or on Wet or Dry Ice
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3103108B Certificate TypeCertificate of Analysis Date27/02/2025 Catalog NumberPV4662
Lot Number2534562D Certificate TypeCertificate of Analysis Date21/02/2025 Catalog NumberPV4662
Lot Number2534562C Certificate TypeCertificate of Analysis Date25/01/2024 Catalog NumberPV4662
Lot Number2399662A Certificate TypeCertificate of Analysis Date09/10/2021 Catalog NumberPV4662
Lot Number1763232B (1) Certificate TypeCertificate of Analysis Date19/07/2017 Catalog NumberPV4662
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ TR-FRET Coregulator Buffer I > 25 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.