missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TRACE™ TR-Wax GC Columns

Analyze a wide range of polar compounds with this low-bleed, general purpose, high polarity GC column.

335.00 € - 1190.74 €

Specifications

Stationary Phase Trace™ TR-WAX
USP Type G16, G20
Polarity Polar
View More Specs

Products 17
Product Code Brand Length (Metric) Diameter (Metric) Inner Film Thickness Price Quantity & Availability  
Product Code Brand Length (Metric) Diameter (Metric) Inner Film Thickness Price Quantity & Availability  
10691386
View Documents
Thermo Scientific™
260W020P
10 m 0.1 mm 0.1 μm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10213033
View Documents
Thermo Scientific™
260W130P
15 m 0.25 mm 0.25 μm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10059142
View Documents
Thermo Scientific™
260W131P
15 m 0.32 mm 0.25 μm
407.10 €
Each
Estimated Shipment: 28-01-2025
to see stock.
Add to basket
 
10170643
View Documents
Thermo Scientific™
260W142P
30 m 0.25 mm 0.25 μm
638.92 €
Each
Estimated Shipment: 24-01-2025
to see stock.
Add to basket
 
10642094
View Documents
Thermo Scientific™
260W143P
30 m 0.32 mm 0.25 μm
680.75 €
Each
Estimated Shipment: 29-01-2025
to see stock.
Add to basket
 
10315253
View Documents
Thermo Scientific™
260W154P
60 m 0.25 mm 0.25 μm
1078.80 €
Each
Estimated Shipment: 30-01-2025
to see stock.
Add to basket
 
10469234
View Documents
Thermo Scientific™
260W155P
60 m 0.32 mm 0.25 μm
1168.13 €
Each
Estimated Shipment: 20-01-2025
to see stock.
Add to basket
 
10120983
View Documents
Thermo Scientific™
260W223P
30 m 0.25 mm 0.5 μm
638.92 €
Each
In Stock
to see stock.
Add to basket
 
10333993
View Documents
Thermo Scientific™
260W224P
30 m 0.32 mm 0.5 μm
680.75 €
Each
Estimated Shipment: 30-01-2025
to see stock.
Add to basket
 
10545475
View Documents
Thermo Scientific™
260W225P
30 m 0.53 mm 0.5 μm
745.45 €
Each
Estimated Shipment: 23-01-2025
to see stock.
Add to basket
 
10743697
View Documents
Thermo Scientific™
260W236P
60 m 0.32 mm 0.5 μm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10458464
View Documents
Thermo Scientific™
260W286P
15 m 0.53 mm 1 μm
432.85 €
Each
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
 
10512075
View Documents
Thermo Scientific™
260W296P
30 m 0.25 mm 1 μm
657.70 €
Each
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
 
10130983
View Documents
Thermo Scientific™
260W297P
30 m 0.32 mm 1 μm
680.75 €
Each
Estimated Shipment: 24-01-2025
to see stock.
Add to basket
 
10796324
View Documents
Thermo Scientific™
260W298P
30 m 0.53 mm 1 μm
736.16 €
Each
Estimated Shipment: 30-01-2025
to see stock.
Add to basket
 
10147834
View Documents
Thermo Scientific™
260W309P
60 m 0.32 mm 1 μm
1190.74 €
Each
Estimated Shipment: 30-01-2025
to see stock.
Add to basket
 
10712517
View Documents
Thermo Scientific™
260W310P
60 m 0.53 mm 1 μm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Analyze a wide range of polar compounds with the Thermo ScientificTM TRACETM TR-Wax GC Column. A polyethylene-glycol-based phase ideal for the analysis of all polar compounds, it is highly cross-linked and fully deactivated to provide excellent peak shape even with short chain alcohols.

Engineered for Superior Performance

  • Polar phase, polyethylene glycol
  • Highly cross-linked and fully deactivated
  • Solvent washable


Applications:

  • Esters
  • Alcohols
  • Ketones
  • Glycols
  • Aromatic isomers


Excellent Quality and Reproducibility:
Recommended Use:

  • Low-bleed even at elevated temperatures
  • Outstanding robustness for difficult separations
  • Reliable and reproducible performance
  • Aldehydes
  • Essential Oils


Alternative Uses:

  • Acids
  • Alcohols
  • Alcohols in beverages
  • Alditol acetates (sugars)
  • Amines–Aliphatic
  • Amines–Aromatic
  • Herbicides
  • Ketones
  • Nitroaromatics
  • Phenols
  • Volatiles


Similar To:

  • DB-Wax
  • BP20
  • Rtx™-Wax
  • Stabilwax™
  • HP20M
  • HP-Wax
  • HP-INNOWax
  • SUPELCOWAX™ 10
  • AT™-Wax
  • Nukol™
  • CP Wax52CB
  • SUPEROX II
  • Carbowax™
  • PE-WAX
  • ZBWAX
Specifications

Specifications

Trace™ TR-WAX
G16, G20
Polar
Videos
SDS
Documents

Documents

Product Certifications

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.