Learn More
Thermo Scientific™ TraceGOLD TG-VRX GC Columns
Thermo Scientific™ TraceGOLD TG-VRX GC Columns provide good resolution and fast analysis times for volatile compounds.
Brand: Thermo Scientific™ 26081-3330
1249.49 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Thermo Scientific™ TraceGOLD TG-VRX GC Columns provide good resolution and fast analysis times for volatile compounds.
- Proprietary stationary phase
- Optimized dimensions for analysis of volatile organic pollutants
- Ideally suited for US EPA Methods 502.2, 524.2, 8021, and 8260
- Similar to DB-VRX, Rtx-VRX
Recommended for:
- Volatile organic pollutants
- Aromatics

Specifications
260°C | |
0.25 mm | |
TraceGOLD™ TG-VRX | |
1.4 μm | |
Non-Polar |
0.25 mm | |
TraceGOLD | |
Volatile Organic Pollutants Analysis, GC/MS for US EPA Method 8021 Compounds | |
60 m | |
60 m x 0.25 mm x 1.40 μm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.