missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trioctylphosphine, 90%, technical grade, AcroSeal™, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
CAS | 4731-53-7 |
Color | White |
Density | 0.8310g/mL |
Boiling Point | 240.0°C (10.0 mmHg) |
Flash Point | 147°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 88% min. (GC) |
Packaging | AcroSeal™ Glass Bottle |
Molecular Formula | C24H51P |
Refractive Index | 1.4670 to 1.4690 (20°C, 589nm) |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0449394 | Certificate of Analysis | 31/01/2025 | 436461000 |
A0449394 | Certificate of Analysis | 17/01/2024 | 436461000 |
A0447031 | Certificate of Analysis | 02/11/2023 | 436461000 |
A0436179 | Certificate of Analysis | 11/01/2023 | 436461000 |
A0401410 | Certificate of Analysis | 14/12/2022 | 436461000 |
Safety and Handling

Product Identifier
- Trioctylphosphine
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : trioctyl-Phosphine
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Trioctylphosphine, 90%, technical grade, AcroSeal™, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction