missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek 500 Labo Coverall
GREENER_CHOICE

Providing an ideal balance of protection, durability and comfort. DuPont™ Tyvek™ CHF7 Hooded Laboratory Coveralls are robust yet lightweight with a 3-piece hood and gusset for improved fit.

11.65 € - 295.00 €

Specifications

Product Type Hooded Coveralls
Material Tyvek
Color White
Waist Style Elastic
Ankle Style Elastic
View More Specs

Products 12
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
15254191
GREENER_CHOICE
View Documents
DuPont™
D14529880
3X-Large
11.95 €
Each
In Stock
to see stock.
Edge
Add to basket
 
11763113
GREENER_CHOICE
View Documents
DuPont™
D14529880
3X-Large
295.00 €
Pack of 25
Estimated Shipment: 30-04-2025
to see stock.
Add to basket
 
15204191
GREENER_CHOICE
View Documents
DuPont™
D14529838
Small
11.65 €
Each
Estimated Shipment: 28-04-2025
to see stock.
Add to basket
 
11753113
GREENER_CHOICE
View Documents
DuPont™
D14529838
Small
289.00 €
Pack of 25
Estimated Shipment: 28-04-2025
to see stock.
Add to basket
 
15224191
GREENER_CHOICE
View Documents
DuPont™
D14529842
Medium
11.65 €
Each
Estimated Shipment: 30-04-2025
to see stock.
Add to basket
 
11743113
GREENER_CHOICE
View Documents
DuPont™
D14529842
Medium
289.00 €
Pack of 25
Estimated Shipment: 25-04-2025
to see stock.
Add to basket
 
11713113
GREENER_CHOICE
View Documents
DuPont™
D14529854
Large
289.00 €
Pack of 25
Estimated Shipment: 25-04-2025
to see stock.
Add to basket
 
15214191
GREENER_CHOICE
View Documents
DuPont™
D14529854
Large
12.12 €
Each
Estimated Shipment: 30-04-2025
to see stock.
Add to basket
 
11733113
GREENER_CHOICE
View Documents
DuPont™
D14529877
2X-Large
289.00 €
Pack of 25
Estimated Shipment: 30-04-2025
to see stock.
Add to basket
 
15244191
GREENER_CHOICE
View Documents
DuPont™
D14529877
2X-Large
11.65 €
Each
Estimated Shipment: 30-04-2025
to see stock.
Add to basket
 
11723113
GREENER_CHOICE
View Documents
DuPont™
D14529869
X-Large
289.00 €
Pack of 25
In Stock
to see stock.
Edge
Add to basket
 
15234191
GREENER_CHOICE
View Documents
DuPont™
D14529869
X-Large
11.65 €
Each
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

  • Excellent barrier against fine particles and fibres
  • Ultra-low-linting and antistatically treated
  • Tyvek zipper and zipper flap for enhanced protection
  • Composed of flash spun high density polyethylene
  • Elasticated face, wrists and ankles as well as glued-in elastic waist
  • Permeable to both air and water vapor, yet repels water-based liquids and aerosols
  • Applications include the pharmaceutical industry, laboratories, cosmetics, optical and electronics
  • Stitched internal seams to help reduce contamination from the inside to the outside of the garment
  • Attached overshoes consisting of garment material with friction-coated sole to provide added slip resistance
Specifications

Specifications

Hooded Coveralls
White
Elastic
Hooded
Stitched, Taped
Zipper flap, tunneled cuff and facial elastics
Tyvek
Elastic
Elastic
3-panel
Category III, Type 5 and 6, EN 1073-2
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DuPont™ Tyvek 500 Labo Coverall

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.